missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCR8 Recombinant Human Monoclonal Antibody (2C10)

Description
CCR8 Recombinant Monoclonal Antibody for Flow
CCR8 encodes a member of the beta chemokine receptor family, which is predicted to be a seven transmembrane protein similar to G protein-coupled receptors. Chemokines and their receptors are important for the migration of various cell types into the inflammatory sites. This receptor protein preferentially expresses in the thymus. I-309, thymus activation-regulated cytokine (TARC) and macrophage inflammatory protein-1 beta (MIP1 beta) have been identified as ligands of this receptor. Studies of this receptor and its ligands suggested its role in regulation of monocyte chemotaxis and thymic cell apoptosis. More specifically, this receptor may contribute to the proper positioning of activated T cells within the antigenic challenge sites and specialized areas of lymphoid tissues. This gene is located at the chemokine receptor gene cluster region. CCR8 expression has been reported in spleen and thymus, natural-killer cells, monocytes, and T cells, and at lower levels in peripheral blood leukocytes.
Specifications
Specifications
| Antigen | CCR8 |
| Applications | Flow Cytometry |
| Classification | Recombinant Monoclonal |
| Clone | 2C10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4 |
| Gene | CCR8 |
| Gene Accession No. | P51685 |
| Gene Alias | C-C; CC chemokine receptor 8; CC chemokine receptor CHEMR1; C-C chemokine receptor type 8; C-C CKR-8; C-C motif chemokine receptor 8; CC-chemokine receptor chemr1; CC-CKR-8; CCR8; CCR-8; cd198; CDw198; chemokine (C-C motif) receptor 8; chemokine (C-C) receptor 8; chemokine (C-C) receptor-like 2; chemokine receptor-like 1; CKR-8; CKRL1; CKR-L1; Cmkbr8; CMKBRL2; CY6; GPRCY6; GPR-CY6; mCCR8; MGC129966; MGC129973; TER1 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?