missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCR9 Antibody (MM0069-6D10), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12136AF488
This item is not returnable.
View return policy
Description
CCR9 Monoclonal specifically detects CCR9 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CCR9 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| P51686 | |
| CCR9 | |
| Human CCR9 transfected 293 cells | |
| 0.1 mL | |
| Cytokine Research, GPCR, Immunology, Innate Immunity, Signal Transduction | |
| 10803 | |
| Store at 4C in the dark. | |
| IgG2 |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0069-6D10 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| C-C chemokine receptor type 9, C-C CKR-9, CC-CKR-9, CCR-9, CDw199, CDw199 antigen, chemokine (C-C motif) receptor 9, G protein-coupled receptor 28, GPR28, GPR-9-6G-protein coupled receptor 28 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction