Learn More
CCT3 Antibody, mFluor Violet 450 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-99267MFV450
Product Code. 18575081
  • € 428.00 / 0.10mL
Description
CCT3 Polyclonal antibody specifically detects CCT3 in Human samples. It is validated for Western Blot, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rabbit
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human CCT3.
7203