missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CCT3 Monoclonal Antibody (12H4)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527872
This item is not returnable.
View return policy
Description
Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human MCF-7 whole cell, human COLO-320 whole cell, human HepG2 whole cell. ICC/IF: A431 cell.
The protein encoded by this gene is a molecular chaperone that is a member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). This complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants have been characterized for this gene. In addition, a pseudogene of this gene has been found on chromosome 8.
Specifications
| CCT3 | |
| Monoclonal | |
| 500 μg/mL | |
| PBS with 4mg trehalose and 0.05mg sodium azide | |
| P49368 | |
| Cct3 | |
| A synthetic peptide corresponding to a sequence at the C-terminus of human CCT3 (497-536aa EPLAVKLQTYKTAVETAVLLLRIDDIVSGHKKKGDDQSRQ). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| 12H4 | |
| Unconjugated | |
| Cct3 | |
| AL024092; CCT3; CCTG; CCT-gamma; chaperonin containing TCP1 subunit 3; chaperonin containing Tcp1, subunit 3 (gamma); chaperonin subunit 3 (gamma); hTRiC5; Matricin; mTRiC-P5; OTTHUMP00000025735; PIG48; T-complex protein 1 subunit gamma; T-complex protein 1, gamma subunit; TCP1 (t-complex-1) ring complex, polypeptide 5; TCP-1-gamma; Tcp1-rs3; TRIC5; TriC-P5 | |
| Mouse | |
| Antigen affinity chromatography | |
| RUO | |
| 7203 | |
| -20°C | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction