Learn More
CCT4 Mouse anti-Human, Clone: OTI4E6, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF810840
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The chaperonin containing TCP1 complex (CCT), also called the TCP1 ring complex, consists of 2 back-to-back rings, each containing 8 unique but homologous subunits, such as CCT4. CCT assists the folding of newly translated polypeptide substrates through multiple rounds of ATP-driven release and rebinding of partially folded intermediate forms. Substrates of CCT include the cytoskeletal proteins actin and tubulin, as well as alpha-transducin.Specifications
| CCT4 | |
| Monoclonal | |
| Unconjugated | |
| CCT4 | |
| CCT-DELTA, Cctd, SRB | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10575 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4E6 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P50991 | |
| CCT4 | |
| Full length human recombit protein of human CCT4 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.