missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCT5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 201.60 - € 552.30
Specifications
| Antigen | CCT5 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, ELISA |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30231565
|
Novus Biologicals
NBP3-38285-100ul |
100 μL |
€ 584.00 € 552.30 / 100µL Save € 31.70 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30228433
|
Novus Biologicals
NBP3-38285-20ul |
20 μL |
€ 214.00 € 201.60 / 20µL Save € 12.40 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CCT5 Polyclonal antibody specifically detects CCT5 in Human,Mouse,Rat samples. It is validated for ELISA,Western BlotSpecifications
| CCT5 | |
| ELISA, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Stem Cell Markers | |
| PBS (pH 7.3), 50% glycerol | |
| 22948 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500 - 1:2000, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| chaperonin containing TCP1, subunit 5 (epsilon), epsilon subunit | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-270 of human CCT5 (NP_036205.1).,, Sequence:, MASMGTLAFDEYGRPFLIIKDQDRKSRLMGLEALKSHIMAAKAVANTMRTSLGPNGLDKMMVDKDGDVTVTNDGATILSMMDVDHQIAKLMVELSKSQDDEIGDGTTGVVVLAGALLEEAEQLLDRGIHPIRIADGYEQAARVAIEHLDKISDSVLVDIKDTEPLIQTAKTTLGSKVVNSCHRQMAEIAVNAVLTVADMERRDVDFELIKVEGKVGGRLEDTKLIKGVIVDKDFSHPQMPKKVEDAKIAILTCPFEPPKPKTKHKLDVTS | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title