Learn More
CCT8L2 Monoclonal Antibody (OTI2A5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA505302S
Encompass_Preferred
 View more versions of this product
Product Code. 17533582
  • € 242.00 / 30µL
Description
CCT8L2 Monoclonal antibody specifically detects CCT8L2 in Human samples. It is validated for Immunocytochemistry, Western Blot
Specifications
Specifications
CCT8L2
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9Y3B7
Mouse
Affinity chromatography
RUO
65003
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Immunocytochemistry, Western Blot
OTI2A5
Unconjugated
MRPL11
CCT8L2; CESK1; chaperonin containing TCP1 subunit 8 like 2; chaperonin containing TCP1, subunit 8 (theta)-like 2; putative T-complex protein 1 subunit theta-like 2; T-complex protein 1 subunit theta-like 2
Full length human recombinant protein of human MRPL11 produced in E.coli.