missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CD 3254
GREENER_CHOICE

Product Code. 18093832
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18093832

Brand: Tocris Bioscience™ 3302/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective RXRα agonist.

Selective RXRα agonist; exhibits no activity at RARα, RARβ or RARγ receptors.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 196961-43-0
Molecular Formula C24H28O3

Specifications

Quantity 50 mg
Formula Weight 364.48
Purity >97%
Molecular Weight (g/mol) 364.485
Chemical Name or Material CD 3254
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.