missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD1 Antibody (76-7-4), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-28221G
This item is not returnable.
View return policy
Description
CD1 Monoclonal specifically detects CD1 in Porcine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
| CD1 | |
| Monoclonal | |
| DyLight 488 | |
| CD1, CD1a antigen, CD1A antigen, a polypeptide, CD1a molecule, cluster of differentiation 1 A, cortical thymocyte antigen CD1A, differentiation antigen CD1-alpha-3, epidermal dendritic cell marker CD1a, FCB6, HTA1, hTa1 thymocyte antigen, R4, T6, T-cell surface antigen T6/Leu-6, T-cell surface glycoprotein CD1a | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Porcine CD1 | |
| Store at 4C in the dark. | |
| IgG2a κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| 76-7-4 | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen | |
| CD1A | |
| Fresh dd miniature swine thymocytes | |
| 0.25 mL | |
| Dendritic Cell Markers, Immunology | |
| 909 | |
| Pig | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction