Learn More
CD100 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 27516-1-AP-20UL
 View more versions of this product
Product Code. 16845675
  • € 145.00 / 20µL
Description
The CD100 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD100. This antibody recognizes human antigen. The CD100 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
CD100
Polyclonal
Unconjugated
20 μL
10507
IgG
Western Blot, Immunohistochemistry, ELISA
267 μg/mL