Learn More
CD100 (Sema4D) Mouse anti-Human, BUV496, Clone: A8, BD Biosciences™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 750140
Product Code. 17251525
  • € 510.00 / 50µg
Description
CD100 (Sema4D) Monoclonal antibody specifically detects CD100 (Sema4D) in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD100 (Sema4D)
Monoclonal
Brilliant Ultraviolet 496
Semaphorin-4D ; SEMA4D; SEM4D; SEMAJ; GR3; C9orf164
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV496 under optimal conditions that minimize unconjugated dye and antibody.
Flow Cytometry
A8
Aqueous buffered solution containing ≤0.09% sodium azide.