missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD105 (Endoglin) Monoclonal Antibody (SN6), Brilliant Ultra Violet™ 661, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
€ 211.00 - € 424.00
Specifications
| Antigen | CD105 (Endoglin) |
|---|---|
| Clone | SN6 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17942061
|
Invitrogen™
376-1057-41 |
25 Tests |
€ 211.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17922011
![]() |
Invitrogen™
376-1057-42 |
100 Tests |
€ 424.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD105 or Endoglin is a Type I transmembrane protein, which is highly expressed on human vascular endothelial cells. It exists on an O- and N-glycosylated homodimer. Up regulation of endoglin expression has been demonstrated in tumor vasculature and proliferating cells, suggesting that it is a proliferation associated endothelial marker. CD105 binds to TGF beta 1 and 3 with high affinity but not to TGF beta 2.Specifications
| CD105 (Endoglin) | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| AI528660; AI662476; CD105; CD105 antigen; cell surface MJ7/18 antigen; Edg; END; Endo; Endoglin; endoglin (Osler-Rendu-Weber syndrome 1); Eng; HHT1; ORW; ORW1; S-endoglin; transmembrane glycoprotein | |
| ENG | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! |
| SN6 | |
| Flow Cytometry | |
| Brilliant Ultraviolet 661 | |
| Mouse | |
| Human | |
| P17813 | |
| 2022 | |
| IgG1 κ | |
| Affinity chromatography | |
| ENG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title