missing translation for 'onlineSavingsMsg'
Learn More

CD105 (Endoglin) Mouse anti-Human, BB700, Clone: 266, BD Horizon™

Mouse Monoclonal Antibody

€ 219.00 - € 447.00

Specifications

Antigen CD105 (Endoglin)
Clone 266
Applications Flow Cytometry
Classification Monoclonal
Conjugate Brilliant Blue 700 (BB700)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15885129
View Documents
BD Biosciences
566529
25 Tests
€ 219.00
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15875129
View Documents
BD Biosciences
566528
100 Tests
€ 447.00
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD105 (Endoglin) Monoclonal antibody specifically detects CD105 (Endoglin) in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD105 (Endoglin)
Flow Cytometry
Brilliant Blue 700 (BB700)
RUO
IgG1 κ
Refrigerate (2 to 8°C)
266
Monoclonal
Mouse
Human
Primary
CD105 (Endoglin)
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.