Learn More
CD106 Mouse anti-Human, Brilliant Violet 650, Clone: 51-10C9, BD Optibuild™
Mouse Monoclonal Antibody
Brand: BD Biosciences 744311
Description
The 51-10C9 monoclonal antibody specifically binds to CD106. CD106 is a 100-110 kDa type I transmembrane sialoglycoprotrein that is also known as Vascular cell adhesion molecule-1 (VCAM-1) and INCAM-110. CD106 is expressed at high levels on the surface of cytokine-stimulated endothelium, and at minimal levels on unstimulated endothelium. VCAM-1 serves as a ligand for the leukocyte integrinsα4β1 (CD49d/CD29 complex; VLA-4) andα4β7 (LPAM-1). The 51-10C9 monoclonal antibody inhibits the in vitro binding of lymphocytes and monocytes to VCAM-1 on stimulated endothelium.
The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
Specifications
| CD106 | |
| 51-10C9 | |
| Brilliant Violet 650 | |
| VCAM-1, Vascular cell adhesion protein 1, INCAM-100, L1CAM | |
| Human VCAM-1 Recombinant Protein | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.