missing translation for 'onlineSavingsMsg'
Learn More

CD106 (VCAM-1) Rat anti-Mouse, Functional Grade, Clone: 429, eBioscience™

Rat Monoclonal Antibody

Brand:  Affymetrix eBioscience 16-1061-81

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15248537

  • € 74.75 / 50µg


Estimated Shipment
11-09-2020
Add to basket

Description

Description

Purity: Greater than 90%, as determined by SDS-PAGE. Endotoxin Level: Less than 0.001 ng/μg antibody, as determined by LAL assay. Aggregation: Less than 10%, as determined by HPLC. This gene is a member of the Ig superfamily and encodes a cell surface sialoglycoprotein expressed by cytokine-activated endothelium. This type I membrane protein mediates leukocyte-endothelial cell adhesion and signal transduction, and may play a role in the development of artherosclerosis and rheumatoid arthritis. Two alternatively spliced transcripts encoding different isoforms have been described for this gene.
Specifications

Specifications

CD106 (VCAM-1)
Monoclonal
1 mg/mL
PBS with no preservative; pH 7.2
P29533
CD106, DKFZp779G2333, INCAM-100, MGC99561, CD106 antigen, vascular cell adhesion protein 1
Rat
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry, Functional Assay, Neutralization
429
Functional Grade
Vcam1
Liquid
Vcam1
IgG2a, kappa
50 μg
4° C
Primary
22329
Videos
Documents

For Research Use Only.