Learn More
CD107a Rat anti-Mouse, FITC, Clone: 1D4B, Proteintech
Rat Monoclonal Antibody
Brand:  Proteintech FITC-65050-500UG
Product Code. 16861166
  • € 228.00 / 500µg
Description
The CD107a antibody from Proteintech is a rat monoclonal antibody generated with plasma membrane fraction of mouse embryo nih/3t3 cell line of mouse CD107a. This antibody recognizes mouse antigen. The CD107a antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Specifications
Specifications
CD107a
Monoclonal
0.5 mg/mL
Rat
Primary
Mouse
Flow Cytometry (Intracellular Staining)
1D4B
FITC
500 μg
16783
IgG2a