Learn More
CD107a Rat anti-Mouse, FITC, Clone: 1D4B, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech FITC-65050-500UG
Description
The CD107a antibody from Proteintech is a rat monoclonal antibody generated with plasma membrane fraction of mouse embryo nih/3t3 cell line of mouse CD107a. This antibody recognizes mouse antigen. The CD107a antibody has been validated for the following applications: FC analysis. This antibody is fitc.
Specifications
| CD107a | |
| Monoclonal | |
| 0.5 mg/mL | |
| Rat | |
| Primary | |
| Mouse |
| Flow Cytometry (Intracellular Staining) | |
| 1D4B | |
| FITC | |
| 500 μg | |
| 16783 | |
| IgG2a |