Learn More
CD107b Rat anti-Mouse, CoraLite 594, Clone: ABL-93, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech CL594-65052-100TESTS
Description
The CD107b antibody from Proteintech is a rat monoclonal antibody to a glycoproteins purified from balb/c mouse embryo 3t3 cell line of mouse CD107b. This antibody recognizes mouse antigen. The CD107b antibody has been validated for the following applications: FC (Intra), IF analysis. This antibody is coralite™594.
Specifications
| CD107b | |
| Monoclonal | |
| 5 μL/Test | |
| Rat | |
| 16784 | |
| IgG2a |
| Immunofluorescence, Flow Cytometry (Intracellular Staining) | |
| ABL-93 | |
| CoraLite 594 | |
| Primary | |
| Mouse | |
| 100 Tests |