missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD107b Rat anti-Mouse, Oligo, Clone: M3/84, BD™ AbSeq ™
Rat Monoclonal Antibody
Brand: BD Biosciences 940483
This item is not returnable.
View return policy
Description
CD107b Monoclonal antibody specifically detects CD107b in Mouse samples. It is validated for Single Cell 3' Sequencing
Specifications
| CD107b | |
| Monoclonal | |
| Oligo | |
| Mac-3; Lamp2; LAMP-2; Lysosome-associated membrane glycoprotein 2; LGP-B | |
| Mouse C57Bl/6 peritoneal exudate cells | |
| RUO | |
| Mouse | |
| IgG1 κ |
| Single Cell 3' Sequencing | |
| Clone M3/84 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Rat | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction