missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD109 Antibody (W7C5), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-75969AF350
This item is not returnable.
View return policy
Description
CD109 Monoclonal specifically detects CD109 in Human samples. It is validated for Flow Cytometry, Flow (Cell Surface).
Specifications
| CD109 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| CD109 | |
| WERI-RB-1 retinoblastoma cell line | |
| 0.1 mL | |
| Primary | |
| The antibody MEM-174 recognizes CD11b antigen (Mac-1 alpha), a 165-170 kDa type I transmembrane protein mainly expressed on monocytes, granulocytes and NK-cells. The CD11b mediates neutrophil and monocyte interactions with stimulated endothelium. | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| W7C5 | |
| Flow Cytometry, Flow (Cell Surface) | |
| 150 kDa TGF-beta-1-binding protein, activated T-cell marker CD109, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7, CD109 antigen, CD109 antigen (Gov platelet alloantigens), CD109 molecule, CPAMD7r150, DKFZp762L1111, FLJ38569, FLJ41966, Gov platelet alloantigens, p180, Platelet-specific Gov antigen, RP11-525G3.1 | |
| Mouse | |
| Protein A purified | |
| Cancer, Cell Biology, Cytokine Research, Immunology, Stem Cell Markers | |
| 135228 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction