missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CD109 Recombinant Rabbit Monoclonal Antibody (2Z2A1)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA551553
This item is not returnable.
View return policy
Description
CD109 Recombinant Monoclonal Antibody for Western Blot, ELISA
CD109 is a member of the alpha2-macroglobulin/complement superfamily. The encoded GPI-linked glycoprotein is found on the cell surface of platelets, activated T-cells, and endothelial cells. The protein binds to and negatively regulates signaling of transforming growth factor beta (TGF-beta).
Specifications
| CD109 | |
| Recombinant Monoclonal | |
| 1.18 mg/mL | |
| PBS with 0.05% BSA, 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
| Q6YHK3 | |
| CD109 | |
| Recombinant protein of human CD109. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
| ELISA, Western Blot | |
| 2Z2A1 | |
| Unconjugated | |
| CD109 | |
| 150 kDa TGF-beta-1-binding protein; 9930012E15Rik; activated T-cell marker CD109; AI480638; C3 and PZP-like alpha-2-macroglobulin domain-containing protein 7; Cd109; CD109 antigen; CD109 molecule; CPAMD7; GARP; Gov platelet alloantigens; GPI-anchored alpha 2 macroglobulin-related protein; GPI-anchored alpha-2 macroglobulin-related protein; p180; platelet-specific Gov antigen; r150; RP11-525G3.1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 135228 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction