missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD115 Rat, FITC, Clone: AFS98, enQuireBio™
Rat Monoclonal Antibody
€ 137.00 - € 326.00
Specifications
Antigen | CD115 |
---|---|
Clone | AFS98 |
Concentration | 0.2mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15952178
|
EnquireBio
QAB53-F-25UG |
25 μg |
€ 137.00
25µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15942178
|
EnquireBio
QAB53-F-100UG |
100 μg |
€ 326.00
100µg |
Estimated Shipment: 25-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
CD115 | |
0.2mg/mL | |
Monoclonal | |
Rat | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
12978 | |
IgG2a κ | |
Protein A/G Purified |
AFS98 | |
Flow Cytometry | |
FITC | |
RUO | |
XP_017173299 | |
Csf1r | |
Primary | |
Store at 2-4°C. |