Learn More
CD116 Mouse anti-Human, BUV563, Clone: hGMCSFR-M1, BD Biosciences™
Mouse Monoclonal Antibody
Brand:  BD Biosciences 749389
Product Code. 17200945
  • ONLINE EXCLUSIVE PRICE
    € 486.00
    € 485.39 / 50µg
    Save € 0.61
    0.13% Off
Description
CD116 Monoclonal antibody specifically detects CD116 in Human samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD116
Monoclonal
Brilliant Ultraviolet 563
CSF2RA; GM-CSF Receptor alpha; GM-CSFRα; GMCSFRA; GMR, SMDP4
Recombinant human GM-CSFR
RUO
Human
IgG1 κ
Flow Cytometry
hGMCSFR-M1
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse
50 μg
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV563 under optimal conditions that minimize unconjugated dye and antibody.