missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CD116 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16392775
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16392775 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16392775 Supplier Invitrogen™ Supplier No. PA585872

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

CD116 Polyclonal Antibody for Flow, ICC/IF, Western Blot

The protein encoded by this gene is the alpha subunit of the heterodimeric receptor for colony stimulating factor 2, a cytokine which controls the production, differentiation, and function of granulocytes and macrophages. The encoded protein is a member of the cytokine family of receptors. This gene is found in the pseudoautosomal region (PAR) of the X and Y chromosomes. Multiple transcript variants encoding different isoforms have been found for this gene, with some of the isoforms being membrane-bound and others being soluble.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD116
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Gene CSF2RA
Gene Accession No. Q00941
Gene Alias CD116; CD116 antigen; CDw116; colony stimulating factor 2 receptor alpha chain; colony stimulating factor 2 receptor alpha subunit; colony stimulating factor 2 receptor, alpha, low-affinity (granulocyte-macrophage); CSF 2R; CSF2R; Csf2ra; CSF2RAX; CSF2RAY; CSF2RX; CSF2RY; Csfgmra; GM CSF R alpha; GM-CSF receptor alpha subunit; GMCSFR; GM-CSFR; GM-CSF-Ra; GM-CSFRalpha; GM-CSF-R-alpha; GMCSFR-alpha; GMR; GMR-alpha; granulocyte-macrophage colony stimulating receptor alpha; granulocyte-macrophage colony-stimulating factor receptor alpha chain; granulocyte-macrophage colony-stimulating factor receptor subunit alpha; MGC3848; MGC4838; SMDP4
Gene Symbols CSF2RA
Host Species Rabbit
Immunogen Synthetic peptide within mouse GM-CSF-R-alpha (CD116) aa 11-60.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 12982
Target Species Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.