missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD117/c-kit Antibody (C117/8399R), Alexa Fluor™ 488, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-20927AF488
This item is not returnable.
View return policy
Description
CD117/c-kit Monoclonal antibody specifically detects CD117/c-kit in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)
Specifications
| CD117/c-kit | |
| Monoclonal | |
| Alexa Fluor 488 | |
| CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein | |
| Recombinant fragment (around aa 50-250) of human CD117/c-kit protein (exact sequence is proprietary) | |
| 0.1 mL | |
| B Cell Development and Differentiation Markers, Cancer, Cancer Stem Cells, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Myeloid derived Suppressor Cell, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers, Tyrosine Kinases | |
| 3815 | |
| Store at 4°C in the dark. | |
| IgG κ |
| ELISA, Immunohistochemistry (Paraffin) | |
| C117/8399R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur