missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD117/c-kit Antibody (YB5.B8), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-43358PE
This item is not returnable.
View return policy
Description
CD117/c-kit Monoclonal specifically detects CD117/c-kit in Human samples. It is validated for Flow Cytometry.
Specifications
| CD117/c-kit | |
| Monoclonal | |
| PE | |
| CD117, CD117 antigen, ckit, C-Kit, EC 2.7.10, EC 2.7.10.1, PBT, piebald trait, Proto-oncogene c-Kit, proto-oncogene tyrosine-protein kinase Kit, SCFRmast/stem cell growth factor receptor, soluble KIT variant 1, Tyrosine-protein kinase Kit, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene homolog, v-kit Hardy-Zuckerman 4 feline sarcoma viral oncogene-like protein | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| YB5.B8 | |
| Flow Cytometry | |
| KIT | |
| Human CD117 | |
| 0.1 mL | |
| Cancer, Cellular Markers, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mast Cell Markers, Mesenchymal Stem Cell Markers, Phospho Specific, Protein Kinase, Signal Transduction, Stem Cell Markers, Tyrosine Kinases | |
| 3815 | |
| Store at 4C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction