Learn More
CD11b Antibody (908649), mFluor Violet 610 SE, Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB11241MFV610
Product Code. 18562146
  • € 695.00 / 0.10mL
Description
CD11b Monoclonal antibody specifically detects CD11b in Mouse samples. It is validated for Western Blot, Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD11b
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
Rat
0.1 mL
Breast Cancer, Cancer, Cell Biology, Cellular Markers, Glia Markers, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Microglia Markers, Myeloid Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers
3684
Store at 4C in the dark.
Western Blot, Flow Cytometry
908649
Western Blot, Flow Cytometry
antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, integrin alpha-M, integrin, alpha M (complement component 3 receptor 3 subunit), Leukocyte adhesion receptor MO1, MAC-1, MAC1A, macrophage antigen alpha polypeptide, MO1A, Neutrophil adherence receptor, neutrophil adherence receptor alpha-M subunit, SLEB6
Chinese hamster ovary cell line CHO-derived recombinant mouse Integrin alpha M beta 2, Phe17-Asn1105 (Integrin alpha M), Gln24-Asn702 (Integrin beta 2), Accession # NP_032427 (Integrin alpha M) and P11835 (Integrin beta 2)
RUO
Primary
Mouse
IgG2b