missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
CD11b Polyclonal specifically detects CD11b in Human, Mouse, Rat, Bovine, C. elegans, Chinese Hamster, Monkey, Rhesus Macaque samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications
Specifications
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin) |
| Dilution | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen |
| Gene Alias | antigen CD11b (p170), CD11 antigen-like family member B, CD11b, CD11b antigen, Cell surface glycoprotein MAC-1 subunit alpha, Complement component 3 receptor 3 subunit, CR-3 alpha chain, CR3A, CR3AMGC117044, integrin alpha-M, integrin, alpha M (complement |
| Gene Symbols | ITGAM |
| Host Species | Rabbit |
| Immunogen | Rabbit Polyclonal CD11b Antibody was made to a synthetic peptide within residues 250-350 of the mouse CD11b protein. [Swiss-Prot# P05555] |
| Quantity | 0.1 mL |
| Regulatory Status | RUO |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?