Learn More
Invitrogen™ CD11b Monoclonal Antibody (ICRF44), Brilliant Ultra Violet™ 661, eBioscience™, Invitrogen™

Mouse Monoclonal Antibody
€ 185.00 - € 375.00
Specifications
| Antigen | CD11b |
|---|---|
| Clone | ICRF44 |
| Concentration | 5 μL/Test |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17737477
|
Invitrogen™
376-0118-41 |
25 Tests |
€ 185.00
Pack of 25 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17747477
![]() |
Invitrogen™
376-0118-42 |
100 Tests |
€ 375.00
Pack of 100 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The ICRF44 monoclonal antibody reacts with human CD11b, a 165 kDa adhesion molecule. CD11b associated with integrin beta2 (CD18) is expressed on the surface of monocytes, granulocytes, activated lymphocytes and a subset of NK cells. CD11b is a receptor for intercellular adhesion molecule family members CD54, CD102 and CD50 as well as for iC3b. These adhesions are crucial in cell-cell and cell-matrix interactions. Applications Reported: This ICRF44 antibody has been reported for use in flow cytometric analysis. Applications Tested: This ICRF44 antibody has been pre-diluted and tested by flow cytometric analysis of normal human peripheral blood cells. This may be used at 5 μL (0.5 μg) per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. Brilliant Ultraviolet™ 661 (BUV661) is a tandem dye that emits at 660 nm and is intended for use on cytometers equipped with an ultraviolet (355 nm) laser. Fixation: Samples can be stored in IC Fixation Buffer (00-8222-49) (100 μL of cell sample + 100 μL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (00-5333-54) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Excitation: 350 nm; Emission: 660 nm; Laser: Ultraviolet Laser.
CD11b Monoclonal antibody specifically detects CD11b in Baboon, Chimpanzee, Cynomolgus Monkey, Human, Rhesus Monkey samples. It is validated for Flow Cytometry.Specifications
| CD11b | |
| 5 μL/Test | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with BSA and 0.09% sodium azide; pH 7.2 | |
| antigen CD11b (p170); antigen CD11b (p170); macrophage antigen alpha polypeptide; CD11 antigen-like family member B; CD11b; CD11B (p170); CD11b/CD18; cell surface glycoprotein MAC-1 alpha subunit; cell surface glycoprotein MAC-1 subunit alpha; complement component 3 receptor 3 subunit; complement component receptor 3 alpha-a; complement receptor type 3; CR3; CR-3 alpha chain; CR3A; F730045J24Rik; integrin alpha M; integrin alpha-M; integrin subunit alpha M; integrin, alpha M; integrin, alpha M (complement component 3 receptor 3 subunit); ITGAM; Leukocyte adhesion receptor MO1; leukocyte integrin alpha-M chain; LOC100351865; Ly-40; MAC1; Mac-1; Mac-1 alpha; Mac-1 alpha (Mac1A); MAC-1 alpha subunit; MAC1A; Mac-1a; macrophage antigen alpha; macrophage antigen alpha polypeptide; MGC117044; MO1A; Neutrophil adherence receptor; neutrophil adherence receptor alpha-M subunit; SLEB6; unnamed protein product | |
| ITGAM | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| ITGAM |
| ICRF44 | |
| Flow Cytometry | |
| Brilliant Ultraviolet 661 | |
| Mouse | |
| Baboon, Chimpanzee, Cynomolgus Monkey, Human, Rhesus Macaque | |
| P11215 | |
| 101866035, 3684, 454069, 714832 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.