Learn More
CD11c Antibody (AP-MAB0806) - Azide and BSA Free, Novus Biologicals™
Armenian Hamster Monoclonal Antibody has been used in 12 publications
Brand:  Novus Biologicals NB110-97871-0.1mg
Product Code. 18778173
  • € 590.00 / 0.10mg
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation
CD11c Monoclonal specifically detects CD11c in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Immunoprecipitation, Immunohistochemistry (Paraffin)
95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, SLEB6
This CD11c antibody was produced from a hybridoma (mouse myeloma fused with spleen cells)from an Armenian Hamster immunized with mouse lymphocytes.
Q9QXH4
Armenian Hamster
0.1 mg