missing translation for 'onlineSavingsMsg'
Learn More

CD11c Monoclonal Antibody (N418), PE-Cyanine5.5, eBioscience™, Invitrogen™

Armenian Hamster Monoclonal Antibody

€ 139.00 - € 342.00

Specifications

Antigen CD11c
Clone N418
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15361020
View Documents Promotion Details
Affymetrix eBioscience
35-0114-80
25 μg
€ 139.00
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 10-04-2024
to see stock.
Please to purchase this item. Need a web account? Register with us today!
15371020
View Documents Promotion Details
Affymetrix eBioscience
35-0114-82
100 μg
€ 342.00
100µg
View alternative products
Save Up to 
»
In Stock
to see stock.
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Description: The N418 monoclonal antibody reacts with mouse CD11c, the integrin alpha X. CD11c non-covalently associates with beta 2 integrin to form the CD11c/CD18 heterodimer. CD11c is expressed by dendritic cells, a subset of Intestinal Intraepithelial Lymphocytes (IEL) and some activated T cells. CD11c/CD18 binds to CD54, iC3b and fibrinogen and plays a role in leukocyte adhesive interactions. N418 binds to CD11c on splenic dendritic cells in the T-dependent areas of mouse spleen and precipitates a 150, 90 kDa heterodimer. Applications Reported: This N418 antibody has been reported for use in flow cytometric analysis. Applications Tested: This N418 antibody has been tested by flow cytometric analysis of mouse splenocytes. This can be used at less than or equal to 0.5 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Light sensitivity: This tandem dye is sensitive photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 μL cell sample + 100 μL IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat.

CD11c Monoclonal antibody specifically detects CD11c in Mouse samples. It is validated for Flow Cytometry.
Specifications

Specifications

CD11c
0.2 mg/mL
Monoclonal
Liquid
RUO
PBS with 0.09% sodium azide; pH 7.2
AI449405; CD11 antigen-like family member C; CD11c; CD11C (p150) alpha polypeptide; complement component 3 receptor 4 subunit; complement receptor 4; Cr4; integrin alpha X; integrin alpha-X; integrin aX; integrin subunit alpha X; integrin, alpha X; integrin, alpha X (antigen CD11C (p150), alpha polypeptide); integrin, alpha X (complement component 3 receptor 4 subunit); Itgax; Leu M5; leu M5, alpha subunit; leukocyte adhesion glycoprotein p150,95 alpha chain; Leukocyte adhesion receptor p150,95; leukocyte surface antigen p150,95, alpha subunit; myeloid membrane antigen, alpha subunit; N418; p150 95 integrin alpha chain; RGD1561123; SLEB6
Itgax
Primary
4° C, store in dark, DO NOT FREEZE!
Itgax
N418
Flow Cytometry
PE-Cyanine5.5
Armenian Hamster
Mouse
Q9QXH4
16411
IgG
Affinity chromatography
Antibody
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.