Learn More
CD11c Armenian Hamster, Biotin, Clone: N418, enQuireBio™
Product Code. 15911698
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
Unit Size:
100µg
25µg
500µg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15911698 25 μg 25µg
15901698 100 μg 100µg
15921698 500 μg 500µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15911698 Supplier EnquireBio Supplier No. QAB23B25ug
Armenian Hamster Monoclonal Antibody
Specifications
Antigen CD11c
Applications Flow Cytometry
Classification Monoclonal
Clone N418
Concentration 0.5mg/mL
Conjugate Biotin
Formulation 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Gene Accession No. NP_067309
Gene Symbols Itgax
Host Species Armenian Hamster
Purification Method Protein A/G Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 16411
Content And Storage Store at 2-4°C.
Isotype IgG