missing translation for 'onlineSavingsMsg'
Learn More

CD11c Hamster anti-Mouse, BV421, Clone: N418, BD Horizon™

Hamster Monoclonal Antibody

€ 142.02 - € 395.91

Specifications

Antigen CD11c
Clone N418
Concentration 0.2mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15824329
View Documents
BD Biosciences
565451
25 μg
ONLINE EXCLUSIVE PRICE
€ 143.00
€ 142.02 / Each
Save € 0.98
0.69% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15834329
View Documents
BD Biosciences
565452
0.1 mg
ONLINE EXCLUSIVE PRICE
€ 396.00
€ 395.91 / Each
Save € 0.09
0.02% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CD11c Monoclonal antibody specifically detects CD11c in Mouse samples. It is validated for Flow cytometry.

CD11c Monoclonal antibody specifically detects CD11c in Mouse samples. It is validated for Flow cytometry.
Specifications

Specifications

CD11c
0.2mg/mL
Monoclonal
Purified
RUO
Mouse
Itgax; Integrin alpha-X; Integrin αX; ITAX; CR4; Complement Receptor 4
IgG2
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BV421 under optimum conditions and unconjugated antibody and free BD Horizon™ BV421 were removed.
N418
Flow Cytometry
Brilliant Violet 421
Hamster
Cell Biology
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse Dendritic Cells
Primary
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.