Learn More
CD11c/Integrin Alpha X Mouse anti-Human, CoraLite 488, Clone: 2F1C10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-60258-100UL
Description
The CD11c/Integrin Alpha X antibody from Proteintech is a mouse monoclonal antibody generated with fusion protein of human CD11c/Integrin Alpha X. This antibody recognizes human antigen. The CD11c/Integrin Alpha X antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
| CD11c/Integrin Alpha X | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 3687 | |
| IgG2a |
| Immunofluorescence | |
| 2F1C10 | |
| Mouse | |
| Primary | |
| Human |