missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c/Integrin alpha X Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 81853-1-RR-20UL
This item is not returnable.
View return policy
Description
The CD11c/Integrin alpha X antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human CD11c/Integrin alpha X. This antibody recognizes human antigen. The CD11c/Integrin alpha X antibody has been validated for the following applications: IHC, IF/ICC, ELISA analysis.Specifications
| CD11c/Integrin alpha X | |
| Recombinant Monoclonal | |
| 750 μg/mL | |
| CD11C; SLEB6 | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| Recombinant Antibody | |
| IgG |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, ELISA | |
| 5O17 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 3687 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction