missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD11c, Rabbit anti-Human,Mouse, Clone: N418, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81066-0.2MG
This item is not returnable.
View return policy
Description
CD11c Monoclonal specifically detects CD11c in Human, Mouse samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| CD11c | |
| Monoclonal | |
| 1 mg/mL | |
| ELISA | |
| 95, 95 alpha chain, 95, alpha subunit, CD11 antigen-like family member C, CD11c, CD11c antigen, CD11Cleu M5, alpha subunit, integrin alpha-X, integrin, alpha X (antigen CD11C (p150), alpha polypeptide), integrin, alpha X (complement component 3 receptor 4 subunit), Leu M5, Leukocyte adhesion glycoprotein p150, Leukocyte adhesion receptor p150, leukocyte surface antigen p150, myeloid membrane antigen, alpha subunit, p150 95 integrin alpha chain, SLEB6 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| The antibody binds to mouse and human CD11c. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG κ |
| ELISA | |
| N418 | |
| Unconjugated | |
| PBS | |
| ITGAX | |
| Hamster spleen cells that had been primed to mouse spleen dendritic cells | |
| 0.2 mg | |
| Dendritic Cell Markers, Signal Transduction | |
| 3687 | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction