missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD122 Rat anti-Mouse, Oligo, Clone: TM-β1, BD™ AbSeq ™
Rat Monoclonal Antibody
Brand: BD Biosciences 940173
This item is not returnable.
View return policy
Description
CD122 Monoclonal antibody specifically detects CD122 in Mouse samples. It is validated for Single Cell 3' Sequencing
Specifications
| CD122 | |
| Monoclonal | |
| Oligo | |
| Il2rb; Il-2Rbeta; IL-2Rβ; IL-15Rbeta; IL-2/15 Receptor-beta; IL-2/15Rbeta | |
| Mouse IL-2Rβ Transfected Cell Line | |
| RUO | |
| Mouse | |
| IgG2b κ |
| Single Cell 3' Sequencing | |
| Clone TM-β1 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Rat | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction