Learn More
CD127 (IL-7Ra) Rat anti-Mouse, APC, Clone: A7R34, Proteintech
Rat Monoclonal Antibody
Brand: Proteintech APC-65093-100UG
Description
The CD127 (IL-7Ra) antibody from Proteintech is a rat monoclonal antibody generated with il-7ra-igg1 fusion protein of mouse CD127 (IL-7Ra). This antibody recognizes mouse antigen. The CD127 (IL-7Ra) antibody has been validated for the following applications: FC analysis. This antibody is apc.
Specifications
| CD127 (IL-7Ra) | |
| Monoclonal | |
| 0.2 mg/mL | |
| Rat | |
| Primary | |
| Mouse |
| Flow Cytometry | |
| A7R34 | |
| APC | |
| 100 μg | |
| 16197 | |
| IgG2a |