Learn More
CD127 / IL-7Ra Rat, APC, Clone: A7R34, enQuireBio™
Rat Monoclonal Antibody
Brand: EnquireBio QAB55-APC-25ug
This item is not returnable.
View return policy
Specifications
| CD127 / IL-7Ra | |
| Monoclonal | |
| 0.5mg/mL | |
| 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2 | |
| Il17ra | |
| Protein A/G Purified | |
| RUO | |
| 16172 | |
| IgG2a κ |
| Flow Cytometry | |
| A7R34 | |
| APC | |
| NP_032385 | |
| Rat | |
| 25 μg | |
| Primary | |
| Store at 2-4°C. |