Learn More
CD127 / IL-7Ra Rat, APC, Clone: A7R34, enQuireBio™
Rat Monoclonal Antibody
Brand:  EnquireBio QAB55-APC-25ug
 View more versions of this product
Product Code. 15902198
  • € 156.45 / 25µg
Specifications
CD127 / IL-7Ra
Monoclonal
0.5mg/mL
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, pH 7.2
Il17ra
Protein A/G Purified
RUO
16172
IgG2a κ
Flow Cytometry
A7R34
APC
NP_032385
Rat
25 μg
Primary
Store at 2-4°C.