missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD13 Mouse anti-Human, BUV496, Clone: L138 (also known as Leu-M7), BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 750631
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD13 Monoclonal antibody specifically detects CD13 in Human samples. It is validated for Flow Cytometry
Especificaciones
| CD13 | |
| Monoclonal | |
| Brilliant Ultraviolet 496 | |
| ANPEP; APN; Aminopeptidase N; Alanyl aminopeptidase; LAP1; PEPN | |
| KG-1a Cell Line | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| L138 (also known as Leu-M7) | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido