missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD132 Rat anti-Human, Oligo, Clone: TUGh4, BD™ AbSeq ™
Rat Monoclonal Antibody
Brand: BD Biosciences 940230
This item is not returnable.
View return policy
Description
CD132 Monoclonal antibody specifically detects CD132 in Human samples. It is validated for Single Cell 3' Sequencing
Specifications
| CD132 | |
| Monoclonal | |
| Oligo | |
| IL-2RG; IL-2Rγ;γc; Commonγ chain; Common gamma chain; gamma c; SCIDX1 | |
| Humanγc Transfected Cell Line | |
| RUO | |
| Human | |
| IgG2b κ |
| Single Cell 3' Sequencing | |
| Clone TUGh4 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Rat | |
| 25 Tests | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography and conjugated to BD AbSeq oligonucleotide under optimal conditions. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction