missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD133 Antibody (170411) [Janelia Fluor™ 585], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB11331JF585
This item is not returnable.
View return policy
Description
CD133 Monoclonal antibody specifically detects CD133 in Human samples. It is validated for Flow Cytometry,CyTOF
Specifications
| CD133 | |
| Monoclonal | |
| Janelia Fluor 585 | |
| AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, prominin (mouse)-like 1, prominin 1 | |
| NS0 mouse myeloma cell line transfected with human CD133, Accession # O43490 | |
| RUO | |
| Primary | |
| Human | |
| Liquid |
| Flow Cytometry, CyTOF | |
| 170411 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.1 mL | |
| Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Cellular Signaling, Core ESC Like Genes, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Lysosome Markers, Neuronal Cell Markers, Neuroscience, Stem Cell Markers | |
| 8842 | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction