Learn More
CD133 Antibody (217106) [FITC], Novus Biologicals™
Rat Monoclonal Antibody
Brand:  Novus Biologicals FAB11332F
Product Code. 18688883
  • € 663.00 / 0.10mL
Description
CD133 Monoclonal antibody specifically detects CD133 in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, CyTOF
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD133
Monoclonal
FITC
PBS
Rat
Protein A or G purified from hybridoma culture supernatant
RUO
Primary
Mouse
Purified
Flow Cytometry, Immunohistochemistry, CyTOF
217106
Flow Cytometry, Immunohistochemistry, CyTOF-ready
AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, prominin (mouse)-like 1, prominin 1
Chinese hamster ovary cell line CHO-derived recombinant mouse CD133, Accession # O54990
0.1 mL
Autophagy, Cancer, Cardiovascular Biology, Cellular Markers, Cellular Signaling, Core ESC Like Genes, Cytokine Research, Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Lysosome Markers, Neuronal Cell Markers, Neuroscience, Stem Cell Markers
8842
Store at 4°C in the dark.
IgG2b