missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD133 Antibody (2F8C5) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 220.00 - € 550.00
Specifications
| Antigen | CD133 |
|---|---|
| Clone | 2F8C5 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready |
| Applications | Western Blot, Flow Cytometry, ELISA, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18265320
|
Novus Biologicals
NBP2-52473 |
0.1 mg |
€ 550.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18641378
|
Novus Biologicals
NBP2-52473-0.025mg |
0.025 mg |
€ 220.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CD133 Monoclonal specifically detects CD133 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, CyTOF-ready.Specifications
| CD133 | |
| 1 mg/mL | |
| Western Blot, Flow Cytometry, ELISA, CyTOF | |
| Unconjugated | |
| Mouse | |
| Cellular Markers, Core ESC Like Genes, Hematopoietic Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers | |
| PBS with 0.05% Sodium Azide | |
| 8842 | |
| Purified recombinant fragment of human PROM1 (AA: 20-108) expressed in E. Coli. | |
| Primary | |
| 97.2 kDa |
| 2F8C5 | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, prominin (mouse)-like 1, prominin 1 | |
| PROM1 | |
| IgG1 | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title