Learn More
CD133 Antibody (3F10), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 4 publications
Brand:  Novus Biologicals NBP2-37741SS
 View more versions of this product
Product Code. 18185519
  • ONLINE EXCLUSIVE PRICE
    € 258.00
    € 243.60 / 25µL
    Save € 14.40
    5% Off
Description
CD133 Monoclonal specifically detects CD133 in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CD133
Monoclonal
Unconjugated
Ascites with 0.03% Sodium Azide
PROM1
Synthesized peptide of human CD133. (20-36)GGQPSSTDAPK AWNYELc
Unpurified
RUO
Primary
Human
IgG1
Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
3F10
Flow Cytometry 1:200 - 1:400, ELISA 1:10000, Immunohistochemistry 1:200 - 1:1000, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:200-1:1000
AC133 Stargardt disease 4 (autosomal dominant), CD133, CD133 retinal 2, prominin (mouse)-like 1, prominin 1
Mouse
133 kDa
0.025 mL
Cellular Markers, Core ESC Like Genes, Hematopoietic Stem Cell Markers, Neuronal Cell Markers, Neuronal Stem Cell Markers, Neuroscience, Stem Cell Markers
8842
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.