Learn More
CD133 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 18495-1-AP-20UL
 View more versions of this product
Product Code. 16818514
  • € 145.00 / 20µL
Description
The CD133 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD133. This antibody recognizes human, mouse antigen. The CD133 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
Specifications
CD133
Polyclonal
Unconjugated
20 μL
8842
IgG
Western Blot, Flow Cytometry, ELISA
300 μg/mL