Learn More
CD133 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 18495-1-AP-20UL
This item is not returnable.
View return policy
Description
The CD133 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human CD133. This antibody recognizes human, mouse antigen. The CD133 antibody has been validated for the following applications: FC, WB, ELISA analysis.
Specifications
| CD133 | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 8842 | |
| IgG |
| Western Blot, Flow Cytometry, ELISA | |
| 300 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |