Learn More
CD134 Mouse anti-Mouse, BUV805, Clone: OX-40, BD Biosciences™
Mouse Monoclonal Antibody
Brand: BD Biosciences 748801
This item is not returnable.
View return policy
Description
CD134 Monoclonal antibody specifically detects CD134 in Mouse samples. It is validated for Flow Cytometry
Specifications
| CD134 | |
| Monoclonal | |
| Brilliant Ultraviolet 805 | |
| OX-40 Antigen; Cd134; Tnfrsf4 | |
| Activated rat lymph node cells | |
| RUO | |
| Mouse | |
| IgG2b κ |
| Flow Cytometry | |
| OX-40 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV805 under optimal conditions that minimize unconjugated dye and antibody. |