Learn More
Invitrogen™ CD137 Recombinant Rabbit Monoclonal Antibody (A24), FITC

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA546631
Description
Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Human 4-1BB (CD137) (Met1-Gln186) and conjugated with FITC under optimum conditions, the unreacted FITC was removed. Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze!.
CD137, also known as TNFRSF9 or 4-1BB, is an inducible costimulatory molecule expressed mainly on activated T cells. Its ligand, known as 4-1BBL, is expressed on activated macrophages, mature B cells, hematopoietic stem cells, and myeloid progenitor cells. CD137 signaling leads to maintaining the survival of activated T cells and CD8+ memory T cells, and clonal expansion of T cells, but also to suppressing myelopoiesis and dendritic cell development. Triggered CD137 induces a cytokine release profile regulating peripheral monocyte survival. Soluble forms of CD137 may provide negative control mechanism for some immune responses.
Specifications
| CD137 | |
| Recombinant Monoclonal | |
| 100 μg/mL | |
| PBS with 0.5% BSA and 0.03% Proclin 300 | |
| Q07011 | |
| TNFRSF9 | |
| Recombinant protein ecoding Human CD137 (Met1-Gln186). | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry | |
| A24 | |
| FITC | |
| TNFRSF9 | |
| 4 1 BB; 41BB; 4-1BB; 4-1BB ligand receptor; A930040I11Rik; AA408498; AI325004; Cd137; CD137 antigen; CDw137; EGK_00183; homolog of mouse 4-1BB; ILA; induced by lymphocyte activation (ILA); interleukin-activated receptor, homolog of mouse Ly63; Ly63; receptor protein 4-1BB; s4 1 BB; s4 1BB; s41BB; sCD137; secreted CD137 antigen; soluble 4 1 BB; soluble 4 1BB; soluble 41BB; soluble CD137; T cell antigen ILA; T-cell antigen 4-1BB; T-cell antigen 4-1BB homolog; T-cell antigen ILA; TNF receptor superfamily member 9; TNFRSF9; Tumor necrosis factor receptor superfamily member 9; tumor necrosis factor receptor superfamily, member 9 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 3604 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.