Learn More
CD138 Mouse anti-Human, PE, Clone: MI15, BD
Mouse Monoclonal Antibody
€ 89.58 - € 251.51
Specifications
| Antigen | CD138 |
|---|---|
| Clone | MI15 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | PE |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15891249
|
BD Biosciences
561704 |
25 Tests |
€ 89.60 € 89.58 / Each Save € 0.02 0.02% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
15816758
|
BD Biosciences
552026 |
100 Tests |
€ 252.00 € 251.51 / Each Save € 0.49 0.19% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MI15 monoclonal antibody specifically binds to CD138 (Syndecan-1), an 85-92 kDa single chain transmembrane protein, strongly expressed on multiple-myeloma-derived cell lines and malignant plasma cell populations. It is also expressed on pre-B cells, immature B cells, and plasma cells, but not on mature circulating B-lymphocytes. Syndecan-1 is a member of the family of transmembrane heparan sulfate proteoglycans. It is also expressed on some non-hematopoietic cells, including embryonic mesenchymal cells, vascular smooth muscle cells, endothelial and neural cells. CD138 binds to many extracellular matrix proteins through its heparan sulfate side-chains, like fibronectin, collagen types I, III, and V, tenascin, thrombospondin, and antithrombin III. It is considered an extracellular matrix receptor that may serve as a co-receptor for fibroblast growth factor and related molecules. Monoclonal antibody MI15 blocks the binding of clone B-B4 but not clone DL-101 (other anti-syndecan-1 antibodies) by flow cytometric analysis.
This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Specifications
| CD138 | |
| Flow Cytometry | |
| PE | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG1 κ | |
| Affinity Purified | |
| Syndecan; SDC; Syndecan 1; SDC1; SYND1 |
| MI15 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human U266 and XG-1 Myeloma Cell Lines | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.