Learn More
Invitrogen™ CD138 Recombinant Rabbit Monoclonal Antibody (017), PerCP

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA546734
Description
Preparation: This antibody was obtained from a rabbit immunized with purified, recombinant Human CD138 (Syndecan-1) (Met1-Glu251) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed. Storage: This antibody can be stored at 2C-8C for twelve months without detectable loss of activity. Protected from prolonged exposure to light. Do not freeze!.
CD138 (syndecan 1) is a transmembrane proteoglycan that can bind a variety of cytokines and modulate their activity, as well as the activity of extracellular matrix components and influence many developmental processes. CD138 is expressed mainly in differentiating keratinocytes and is transiently upregulated in all layers of the epidermis upon tissue injury. It is also highly expressed on plasma cells and can be detected even on fibroblasts, vascular smooth muscle cells and endothelial cells. Up-regulation and down-regulation of CD138 on the cell surface often correlates with the gain of cancerous characteristics. Serum levels of the shedded soluble sCD138 are used as a prognostic factor of cancerogenesis.
Specifications
| CD138 | |
| Recombinant Monoclonal | |
| 100 μg/mL | |
| PBS with 0.5% BSA and 0.03% Proclin 300 | |
| P18827 | |
| Sdc1 | |
| Recombinant protein ecoding Human CD138 (Met1-Glu251). | |
| 100 Tests | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry | |
| 017 | |
| PerCP | |
| Sdc1 | |
| AA408134; AA409076; CD antigen 138; CD138; CD138 antigen; heparan sulfate proteoglycan fibroblast growth factor receptor; HSPG; sCD138; SDC; Sdc1; soluble CD138; Sstn; syn-1; Synd; SYND1; Synd-1; SYNDECA; syndecan; syndecan 1; syndecan proteoglycan 1; Syndecan1; syndecan-1; synstatin | |
| Rabbit | |
| Protein A | |
| RUO | |
| 6382 | |
| 4°C, store in dark, DO NOT FREEZE! | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.