missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD14 Antibody (503), Alexa Fluor™ 405, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90362AF405
This item is not returnable.
View return policy
Description
CD14 Monoclonal specifically detects CD14 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Product Image.
Specifications
| CD14 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence | |
| 503 | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence | |
| CD14 antigen, CD14 molecule, monocyte differentiation antigen CD14, Myeloid cell-specific leucine-rich glycoprotein | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Mouse CD14 (NP_033971.1; Met1-Pro345). | |
| 0.1 mL | |
| Adaptive Immunity, Apoptosis, Asthma, Biologically Active Proteins, Cancer, Cell Biology, Cytokine Research, Immunology, Innate Immunity, Myeloid Cell Markers, Stem Cell Markers | |
| 929 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction